missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scie Plas TV400 Standard Twin-Plate Maxi-Gel Vertical Electrophoresis Unit
Designed specifically for higher resolution PAGE techniques requiring self-cast gels.
Brand: Scie Plas TV400
This item is not returnable.
View return policy
Description
- Twin-plate 20.5 x 20cm maxi-gel unit
- Large format with 20.5 x 20cm glass plate dimensions are ideal for running higher percentage polyacrylamide gels to resolve smaller proteins of similar MW
- Float glass plates of 4mm thick provides uniform gel thickness and sample migration, and with ground edges inhibits sample leakage during casting
- Triple screws present on each gel-clamp acts in conjunction with a counterbalanced, dovetailed silicone gasket to disperse pressure evenly along the height and breadth of the gel as the screws are tightened
Specifications
2.5 to 3.5 hr. | |
W x H: 16.5 x 17.5 cm | |
Plate: 0.4 cm | |
Gel Tank: 4200 mL | |
W x H: 28.5 x 27.5 cm | |
15 cm | |
20 to 30 mA |
TV400 Standard Twin-Plate Maxi-Gel Vertical Electrophoresis Unit | |
Twin-plate wide format mini-gel unit with GRM and gel tank, lid, 2 x (20.5 x 20cm; W x H) plain glass plates, 4 x 1mm spacers, 2 x spacer aligners, 2 x (20.5 x 20cm) notched glass plates, 1 x dummy plate and 2 x 1mm thick 24-sample combs | |
0.1 cm | |
Inner Buffer Chamber: 700 mL | |
275 mm | |
175 to 225 V |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction