Learn More
Eppendorf™ twin.tec™ PCR Plates BioBased
Discover the new twin.tec™ PCR Plates BioBased made of bio-based polymer (mass balance approach). These plates allow you to significantly reduce your consumable-related carbon footprint while maintaining identical technical performance.
Brand: Eppendorf™ 0030129857
Additional Details : Weight : 0.75400kg
Description
- Reduction of consumable-related carbon footprint in the lab
- Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
- Unique laser-engraved optical guiding grid and OptiTrack matrix for quick orientation when pipetting manually
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
- Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
- Raised well rims for effective sealing, also reduces risk of cross-contamination
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
- Available with barcode (upon request)
Specifications
Custom Barcoding Available | |
8 x 12 | |
Green | |
Skirted | |
150 μL | |
Non-sterile | |
Conical Bottom (V-well) | |
PCR Ready |
SBS Standard | |
Eppendorf | |
BioBased PCR Plates | |
Polypropylene, Polycarbonate | |
96 | |
Non-treated | |
25 Pk. |