Learn More
DuPont™ Tychem 2000 C
Bright yellow color provides high visibility. Lightweight and durable fabric and a polymeric barrier coating offers good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure).
273.00 SEK - 6440.00 SEK
Specifications
Product Type | Hooded Coveralls |
---|---|
Material | Tychem |
Color | Yellow |
Waist Style | Elastic |
Ankle Style | Elastic |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
12658341
|
DuPont™
D13494969 |
Medium |
6440.00 SEK
Pack of 25 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
12369419
|
DuPont™
D13494969 |
Medium |
273.00 SEK
Each |
In Stock
Log in to see stock. |
|||||
12379419
|
DuPont™
D13494990 |
Small |
273.00 SEK
Each |
In Stock
Log in to see stock. |
|||||
12648341
|
DuPont™
D13494990 |
Small |
6440.00 SEK
Pack of 25 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11743093
|
DuPont™
D13395699 |
X-Large |
6440.00 SEK
Pack of 25 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
12389419
|
DuPont™
D13395699 |
X-Large |
273.00 SEK
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
12359419
|
DuPont™
D13395589 |
Large |
273.00 SEK
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
11733093
|
DuPont™
D13395589 |
Large |
6440.00 SEK
Pack of 25 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
12932159
|
DuPont™
D13395560 |
2X-Large |
273.00 SEK
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11753093
|
DuPont™
D13395560 |
2X-Large |
6440.00 SEK
Pack of 25 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
Description
- Hooded coverall with elastic face opening
- Adhesive chin flap for tight fit around respirator
- Stitched and over-taped seams
- Souble self-adhesive zipper flap
- Elastic cuffs, ankles and waist
- Thumb loops prevent sleeves from riding up
- 100% particle tight fabric
- Excellent barrier against a broad range of concentrated inorganic chemicals and biological hazards
- Antistatic treatment (inside)
Specifications
Hooded Coveralls | |
Yellow | |
Elastic | |
Elastic | |
Stitched, Taped | |
EN 14126, EN 1073-2, EN 1149-5 |
Tychem | |
Elastic | |
Thumb Loop | |
Double Zipper with Self-adhesive Flap | |
Category III, Type 3-B, 4-B, 5-B and 6-B |