Learn More
DuPont™ Tychem™ 6000F Hooded Coverall with FaceSeal
1144.17 SEK valid until 2024-12-31
Use promo code "22918" to get your promotional price.
Use promo code "22918" to get your promotional price.
Click to view available options
Type: Coverall
Description
- Elasticated waist for an optimal fit
- Double cuffs with attached undergloves
- Dissipative socks with boot flap and back entry for enhanced wearer protection
- Lightweight and durable fabric
- Offers excellent chemical permeation resistance to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Certifications/Compliance | (EU) 2016/425, EN 14126, EN 1149-5 |
Color | Gray |
Size | X-Large |
Closure Type | Zipper |
Includes | Attached dissipative socks and boot flaps |
Product Type | Coverall |
Seam Type | Stitched and Taped |