Learn More
MedChemExpress Tyrosine Protein Kinase JAK 2 (Phospho-Tyr8, 9)
MedChemExpress Tyrosine Protein Kinase JAK 2 (Phospho-Tyr8, 9) is a peptide corresponding to amino acids 475 to 491 of mouse JAK2.
Brand: MedChemExpress HY-P1590
This item is not returnable.
View return policy
Specifications
247171-44-4 | |
COVID-19-immunoregulation | |
Peptides | |
Room temperature |
Please store the product under the recommended conditions in the Certificate of Analysis. | |
2082.1 g/mol | |
1 mg |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.