missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ Hooded Coverall 800 J, Model TJ 198 Ta

Product Code. 15591182
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Achieve an effective barrier against low-concentrated water-based inorganic chemicals and small-sized hazardous particulates, as well as oil repellency using Tyvek™ Impervious Technology. DuPont™ Tyvek™ Hooded Coverall 800 J, Model TJ 198 Ta provides an ideal balance of protection, durability and comfort.

  • Robust yet lightweight (<300g)
  • Self-adhesive zipper flap
  • Self-adhesive chin flap for tight seal of suit to mask
  • Elasticated face, wrists and ankles as well as glued-in waist elastic
  • Thumb loops to prevent sleeves from riding up
  • Available in white, sizes S to XXXL
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hooded Coveralls
Size Medium
Material Tyvek
Color White
Disposable Single-use
Waist Style Elastic (Glued-in)
Ankle Style Elastic
Cuff Style Elastic, Thumb Loop
Neck Style Hooded
Closure Type Zipper with Self-adhesive Flap and Chin Flap
Seam Type Stitched, Taped
Certifications/Compliance Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B, EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), Antistatic treatment (EN 1149-5) - on inside
Size (Metric) Chest 92 to 100 cm
For Use With (Application) Very humid applications requiring chemical, liquid protection and/or oil repellency
Quantity 25/Pk.
Show More Show Less

The user should make the risk assessment to determine the protection required.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'