Learn More
DuPont™ Tyvek 500 Hood
Wide flange and elastic around neck and face offer enhanced protection for body parts that are more exposed to hazardous substances.
Brand: DuPont™ D13395804
1549.15 SEK valid until 2024-12-31
Use promo code "22918" to get your promotional price.
Additional Details : Weight : 2.90000kg
Description
- Flash-spun HDPE construction
- Wide flange and elastic around neck and face
- Stitched internal seams
- White
- PPE Category I
- Antistatic treatment (EN 1149-5) - on both sides
Specifications
Tyvek Hood | |
HDPE | |
Stitched | |
100/Pk. |
Universal | |
White | |
PPE Category I, EN 1149-5 | |
Flashspun HDPE |