missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ULK1 |
---|---|
Concentration | 0.35 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850575
|
Proteintech
20986-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16840575
|
Proteintech
20986-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ULK1, also known as ATG1, is a key serine/threonine protein kinase probably acting at the most upstream step of autophagosome formation. Knockout of ULK1 results in a severe defect in the autophagy pathway. ULK1 is highly conserved among eukaryotes, and are the Unc-51-like kinases, ULK1 and ULK2 in mammals. ULK1 is ubiquitously expressed and involved in autophagy in response to starvation. It is the target of the TOR kinase signaling pathway that regulates autophagy through the control of phosphorylation status of ATG13. ULK1 also plays a role early in neuronal differentiation.Specifications
ULK1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat | |
O75385 | |
360827, 8408 | |
Peptide | |
Primary | |
-20°C | |
ULK1 |
0.35 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ATG1, KIAA0722, ULK1, Unc 51 like kinase 1, UNC51, Unc51.1 | |
Ulk1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |