Learn More
Hughes Unheated Emergency Safety Shower
Floor mounted unheated emergency safety shower, suitable for use in all applications.
Brand: Hughes HSHS0018GS
This item is not returnable.
View return policy
Description
- Floor mounted unheated emergency safety shower
- ANSI compliant model ideal for use in indoor applications not subject to the possibility of freezing or overheating
- Delivers the ANSI recommended 76 liters per minute of potable water
- Available with either galvanised powder coated mild steel (G) or stainless steel (GS) pipework
Specifications
Stainless Steel | |
Floor | |
Safety Shower |