Learn More
Fritsch™ Universal cutting mill Pulverisette 19 throughput up to 60kg/hr 230V 50/60Hz 2200W Fritsch
Brand:  Fritsch™ 19.1020.00
Encompass
Product Code. 13108123
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
All grinding parts can be easily removed for cleaning. Different rotors, various knife geometries and replaceable blades ensure maximum flexibility and durability. For controlling the abrasion properties, the grinding tools are offered in different materials. The optimum airflow ensures fast grinding and secure protection against clogging. The patented Fritsch sample exhauster with cyclone separator (optional) ensures simple feeding, fast throughput and good cooling.
Please note that Pulverisette 15 and Pulverisette 19 are intended for sample processing only, not for continuous or heavy duty industrial use.
Ideal for pre- and fine grinding of soft to medium hard as well as fibrous or tough materials. Essential for the areas of plastics, textiles, agriculture and forestry, environmental, construction materials, chemistry, foodstuffs and for sample preparation for RoHS analysis.
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Fritsch™ Universal cutting mill Pulverisette 19 throughput up to 60kg/hr 230V 50/60Hz 2200W Fritsch