Learn More
Kern Sohn Unlined Wooden Box for Single Weight
Wooden case for a single weight.
199.87 SEK - 6936.57 SEK
Specifications
Description | Unlined |
---|---|
Material | Wood |
For Use With (Equipment) | Class F2 and M1 Weights |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
17302153
|
Kern Sohn
337-010-200 |
1 g weight |
246.89 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
17322153
|
Kern Sohn
337-020-200 |
2 g weight |
199.87 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
17342153
|
Kern Sohn
337-030-200 |
5 g weight |
211.62 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
17362153
|
Kern Sohn
337-040-200 |
10 g weight |
211.62 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
17382153
|
Kern Sohn
337-050-200 |
20 g weight |
211.62 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
17302163
|
Kern Sohn
337-060-200 |
50 g weight |
235.14 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
17322163
|
Kern Sohn
337-070-200 |
100 g weight |
258.65 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
17332163
|
Kern Sohn
337-080-200 |
200 g weight |
282.16 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
17352163
|
Kern Sohn
337-090-200 |
500 g weight |
329.19 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
17372163
|
Kern Sohn
337-110-200 |
1 kg weight |
505.54 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
17392163
|
Kern Sohn
337-120-200 |
2 kg weight |
482.03 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
17312173
|
Kern Sohn
337-130-200 |
5 kg weight |
999.33 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
17332173
|
Kern Sohn
337-140-200 |
10 kg weight |
1093.39 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
17382173
|
Kern Sohn
337-150-200 |
20 kg weight |
4526.40 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
17332183
|
Kern Sohn
337-160-200 |
50 kg weight |
6936.57 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
Description
- Wooden
- For button/compact weight design
Spécification
Unlined | |
Wood | |
Class F2 and M1 Weights |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.
Nom du produit