Learn More
Poulten Graf™ Unplugged Pasteur Pipets
Conforms to BS 5732
Brand:  Poulten Graf™ D814/PP
Product Code. 10025901
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Nominal rate of 30 drops/mL to 35 drops/mL. BS 5732.
Specifications
Specifications
Glass
270 mm
Pasteur Pipet
Single-use