Learn More
Kartell™ Plastilab™ L-Shaped Untapered Connectors
Designed for use with hose system and vacuum lines.
Brand:  Kartell™ 0125400
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15631825
  • 65.90 SEK / Pack of 100
Estimated Shipment: 16-08-2024
to see stock.
Add to basket
Description
Description
  • Made from polypropylene
  • Autoclavable
  • Nominal dimensions
  • Compliance
    Meet Food and Agriculture regulations
    Specifications
    Specifications
    Untapered L Tubing Connector
    12 mm
    8 mm bore
    CE marked, IVD
    100/Pk.
    L-shaped