Learn More
Kartell™ Plastilab Untapered T-Shaped Tubing Connectors
BIOPHARMA
Designed with serrations to hold hoses tightly and prevent slipping.
Brand:  Kartell™ 0046100
740.05 SEK valid until 2024-12-31
Use promo code "21793" to get your promotional price.
 View more versions of this product
Product Code. 12519666
  • 779.00 SEK / Pack of 100
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Description
Designed with serrations to hold hoses tightly and prevent slipping.
  • Made from polypropylene
  • Autoclavable
  • Chemical- and temperature-resistant
Compliance
Meet Food and Agriculture regulations
Specifications
Specifications
Meet with the Food and Drug regulations. 90/128/CEE 92/39/CEE e FDA 21 CFR 177.1520, CE and IVD
Autoclavable
Suitable for foodstuff
4.4mm bore
Untapered T Tubing Connector
Polypropylene
100/Pk.
T-shape