Learn More
Kinesis™ Peristaltic Tubing Adapters
Connect 1/16 in. OD rigid/semi-rigid tubing to soft-wall tubing
Brand: Kinesis™ 02014-39
This item is not returnable.
View return policy
Description
Peristaltic tubing adapters feature specially designed nose to allow soft tubing to fit over the nose then be held in place by the retainer sleeve. To connect peristaltic tubing to rigid tubing with a different OD, order a flangeless fitting with 1/4-28 flat-bottom configuration.Specifications
Peristaltic Tubing Adapter | |
0.048 to 0.110 in. | |
Peristaltic to 1/16 in. O.D. Rigid |