Learn More
Idex Fingertight Two-Piece Fitting, Long Thread, Natural PEEK, 1/16 in. OD Tubing, 10-32 Coned
Popular threaded coned port configurations support quality high-pressure connections
Brand: Idex F-330X
This item is not returnable.
View return policy
Description
Popular threaded coned port configurations support quality high-pressure connections
- Perfect for analytical instruments, in-vitro diagnostics, electronics and other biotechnology applications
Two-piece fittings comprise of a separate nut and ferrule. Save money by replacing only the ferrule. For mass spectrometer electrospray application order ferrule 02019-15.
Specifications
Fingertight Two-Piece Fitting | |
PEEK | |
1/16 in. OD Tubing |
Ferrule | |
Natural |