Learn More
Idex Threaded Port Adapters
Threaded port adapters help bring products with different thread configurations together
590.00 SEK
Product Code | Brand | Diameter (English) Inner | Material | Thread Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Inner | Material | Thread Size | Price | Quantity & Availability | |||
12977729
|
Idex
P-626 |
0.02 in. | PEEK | 10-32 Coned to M6 Flat Bottom |
590.00 SEK
Each |
Estimated Shipment: 17-12-2024
Log in to see stock. |
|||
Specifications
1000 psi |