Learn More
Fisherbrand™ Upright ULT Freezer Racks for Microtiter Plates
Fisherbrand inventory control racks for 96-Well and 384-Well microtiter plates. They fit all standard types of upright ULT freezers.
1210.00 SEK - 1520.00 SEK
Specifications
Product Type | Side Access Freezer Rack |
---|---|
Material | Stainless Steel |
For Use With (Application) | 96/384-Well Microtiter Plates |
For Use With (Equipment) | Upright ULT Freezer |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15729559
|
Fisherbrand™
|
Plates with Lid (70) |
1210.00 SEK
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
15739559
|
Fisherbrand™
|
Plates with Lid (84) |
1400.00 SEK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15749559
|
Fisherbrand™
|
Plates with Lid (96) |
1520.00 SEK
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
Description
- Holds up to 96 plates with lid or 120 plates without lid
- Made of high quality stainless steel
- Side opening with handles on the front for easy plate retrieval.
Specifications
Side Access Freezer Rack | |
Stainless Steel | |
96/384-Well Microtiter Plates | |
Upright ULT Freezer |