missing translation for 'onlineSavingsMsg'
Learn More
Learn More
URIDINE-5'-DIPHOSPHATE, MP Biomedicals
Click to view available options
Quantity:
25 mg
100 mg
500 mg
1 g
Specifications
Specifications
Quantity | 25 mg |
Formula Weight | 448.17 (Anhydrous) |
Chemical Name or Material | URIDINE-5'-DIPHOSPHATE, MP Biomedicals |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction