Learn More
Uvex™ 1 Safety Shoe S2
Sporty, multifunctional safety shoe
1595.00 SEK
Specifications
Certifications/Compliance | ESD certified, EN ISO 20345: 2011 S2 SRC |
---|---|
Color | Black |
Design | Loafer |
Material | Microvelour |
Toe Material | Composite |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15381203
|
Uvex™
8544836 |
36 |
1595.00 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
15391203
|
Uvex™
8544837 |
37 |
1595.00 SEK
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
15301213
|
Uvex™
8544838 |
38 |
1595.00 SEK
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||||
15311213
|
Uvex™
8544839 |
39 |
1595.00 SEK
Each |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
15321213
|
Uvex™
8544840 |
40 |
1595.00 SEK
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
15331213
|
Uvex™
8544841 |
41 |
1595.00 SEK
Each |
In Stock
Log in to see stock. |
|||||
15341213
|
Uvex™
8544842 |
42 |
1595.00 SEK
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
15351213
|
Uvex™
8544843 |
43 |
1595.00 SEK
Each |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||||
15361213
|
Uvex™
8544844 |
44 |
1595.00 SEK
Each |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
15371213
|
Uvex™
8544845 |
45 |
1595.00 SEK
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
15381213
|
Uvex™
8544846 |
46 |
1595.00 SEK
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||||
15391213
|
Uvex™
8544847 |
47 |
1595.00 SEK
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
15301223
|
Uvex™
8544848 |
48 |
1595.00 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
15311223
|
Uvex™
8544849 |
49 |
1595.00 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
Description
Specifications
ESD certified, EN ISO 20345: 2011 S2 SRC | |
Loafer | |
Composite | |
Unisex | |
No |
Black | |
Microvelour | |
11 mm | |
Trainers | |
ESD Safety Shoe |