Learn More
Brady™ V1100 Kit for V4500 Barcode Reader EU
Ensure uninterrupted productivity with the V1100 Kit for V4500 Barcode Scanner - complete with spare charger, batteries and essential adapters for seamless scanning anytime, anywhere.
Brand: Brady™ 322621
Additional Details : Weight : 0.56000kg Transport : UN number : 3480 Chem class : 9
Description
- Charge up to two batteries at once with the spare battery charger
- Battery lasts a full shift with over 75,000 scans
- All kit components for use with the V4500 Barcode Reader
- This kit includes a spare charger, two batteries, an AC adapter and a EU power cord
Specifications
Battery Charger | |
Charging over battery charger (0 to 100%) 4 hr. | |
CE, UKCA | |
5% to 95% (non-condensing) | |
132 x 85 x 65 mm |
0.56 kg | |
1.2 lb. | |
Asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, general identification, inventory and inspection labelling, warehouse marking | |
Two V1200 Batteries, an AC Adapter, a EU Power Cable, and a V1100 Battery Charger | |
3 Years |