Learn More
BD Vacutainer™ Crossmatch Serum Tubes
Available with spray-coated EDTA or silica clot act additive. BD Vacutainer™ Crossmatch Serum Tubes are plastic tubes with a pink Hemogard™ closure and extra-large block label that are used to obtain either a serum sample or a whole blood or EDTA plasma sample.
Brand: BD 367941
This item is not returnable.
View return policy
Description
The BD Vacutainer™ system is a closed system optimally designed and tested for controlling pre-analytical variability throughout the specimen collection and handling process. The BD Vacutainer™ Crossmatch tube is identified by:- Sterile evacuated tube made of durable PET plastic
- Color coded pink Hemogard™ Cap designed for use with Vacutainer needles and tube holders
- Specialized label is larger than a typical block label to allow more information to be recorded
- Mixing: Tubes should be gently inverted 180° and back several times (5-6X for Silica, 8-10X for EDTA)
- Centrifugation conditions: ≤1300 g for 10 minutes at room temperature (25°C)
Specifications
Vacutainer Crossmatch Tube | |
PET | |
Hemogard | |
Vacutainer™ Needle and Tube Holder | |
13 x 100 mm | |
Crossmatch (Extra Large Block) |
K2 EDTA (Spray) | |
Sterile | |
6 mL | |
Pink | |
Used to obtain either a serum sample or a whole blood or EDTA plasma sample |