Learn More
VACUUBRAND™ VACUU·LAN™ Manual Control Module With A5 Mounting Base For VCL AR Hoods
The first step is to select the right mounting base. They are used for mounting on or in the wall and they form the link between the vacuum network and vacuum connection;The mounting base A5 is designed for installation of vacuum networks into laboratory furniture. The network tubing is laid hidden behind walls or underneath the work plate. The vacuum ports are mounted from the front.
Brand: VACUUBRAND™ 20677195
This item is not returnable.
View return policy
Description
Specifications
For VCL AR Hoods | |
1 Each | |
Manual Control Module |