Learn More
Memmert™ Vacuum Oven Thermoshelf
Brand: Memmert™ VO49B00743
Additional Details : Weight : 1.00000kg
Specifications
Calibration certificate | |
Shelf | |
Local Temperature Sensing (Pt100, 4-Wire-Circuit) |
Model 400 | |
Individual overtemperature protection for each shelf MLOP (Multi-Level-Overtemperature-Control) | |
Aluminum |