Learn More
BINDER™ Pump Chamber with VAP 5 Vacuum Pump
Pump Chamber with VAP 5 Vacuum Pump - 230 V/50-60 Hz, for mounting vacuum pumps including chemical membrane pump VAP 5 with nominal air flow 3.6 m³;/hr., final pressure 3 mbar, with separator and emission condenser, suitable for flammable solvents and vacuum pump connection kit (ATEX).
Brand: BINDER™ 8012-2061
This item is not returnable.
View return policy
Description
Pump chamber with VAP 5 vacuum pump for use with vacuum pumps with chemical membranes.
- Nominal air flow
- With separator and emission condenser
- Suitable for flammable solvents
- Includes vacuum pump connection kit (ATEX)
Specifications
1010 mbar | |
3.6 m3/hr. | |
Vacuum Pump | |
3 Mbar | |
230 V |
Pump chamber with vacuum pump VAP 5 - 230 V / 50-60 Hz option model, Pump chamber for mounting vacuum pumps including chemical membrane pump VAP 5 with nominal air flow 3.6 m3/hour, final pressure 3 Mbar, with separator and emission condenser, suitable for flammable solvents and vacuum pump connection kit (ATEX). | |
50/60 Hz | |
IP55 | |
BINDER Chambers VDL 56 Vacuum Drying Chamber |