Learn More
Lenz Laborglasintrumente Vacuum Stopcocks, 1-Way, Straight
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 02902408
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10151661
  • 911.60 SEK / Each
Estimated Shipment: 14-10-2024
to see stock.
Add to basket
Description
Description
  • With straight side arms and hollow glass plug
  • Material: borosilicate glass 3.3
  • Leakage rate acc. to DIN 12540-1
Specifications
Specifications
8 mm
Borosilicate Glass (3.3)