Learn More
Valeric acid, 99%, Thermo Scientific Chemicals Product Code.: 11483347
Valeric acid, 99%, Thermo Scientific Chemicals
Product Code. 11483347
500 mL
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Product Code. 11483347
Brand: Thermo Scientific Alfa Aesar A16238.AP
Request Bulk or Custom Format
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Valeric acid, is used as a sex attractant of the sugar beet wireworm, Limonius californicus. It is used predominantly as an intermediate in the manufacture of flavors and perfumes, ester type lubricants, plasticizers and vinyl stabilizesrs. It is a food additive used as a synthetic flavoring substance dan adjuvant. It is an inert ingredient in controlling pest.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Valeric acid, is used as a sex attractant of the sugar beet wireworm, Limonius californicus. It is used predominantly as an intermediate in the manufacture of flavors and perfumes, ester type lubricants, plasticizers and vinyl stabilizesrs. It is a food additive used as a synthetic flavoring substance dan adjuvant. It is an inert ingredient in controlling pest.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 109-52-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00004413
InChI Key NQPDZGIKBAWPEJ-UHFFFAOYSA-N
Synonym valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, nShow MoreShow Less
PubChem CID 7991
ChEBI CHEBI:17418
IUPAC Name pentanoic acid
SMILES CCCCC(=O)O
Spécification
Melting Point -34°C
Boiling Point 184°C to 186°C
Linear Formula CH3(CH2)3CO2H
UN Number UN3265
Beilstein 969454
Merck Index 14,9904
Solubility Information It is soluble in water.
Formula Weight 102.13
Percent Purity 99%
Density 0.938
Quantity 500 mL
Flash Point 88°C (190°F)
Odor Unpleasant
Refractive Index 1.408
Chemical Name or Material Valeric acid
Afficher plus Afficher moins
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465713 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog NumberA16238.AE,A16238.0F,A16238.AP
Lot NumberA0440264 Certificate TypeCertificate of Analysis Date12/03/2024 Catalog NumberA16238.AE,A16238.0F,A16238.AP
Lot NumberA0435809 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberA16238.AE,A16238.0F,A16238.AP
Lot Number10238820 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA16238.AE,A16238.0F,A16238.AP
Lot NumberK15Y005 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16238.AE,A16238.0F,A16238.AP
compliance-icons compliance-icons
Identifiant produit
  • Valeric acid
Mention d'avertissement
  • Danger
Catégorie de danger
  • Skin corrosion/irritation Category 1 B
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Mention de danger
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Conseils de prudence
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Valeric acid, 99%, Thermo Scientific Chemicals >
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis