Learn More
Eppendorf™ Valve, For Varitip S
GREENER_CHOICE
Tailored to different vessels. Choose from a variety of Eppendorf Varitips to meet your needs.
Brand:  Eppendorf™ 0030050541
890.00 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10538911
  • 893.56 SEK / Pack of 100
In Stock
to see stock.
Explore more special offers
Description
  • Eppendorf Varitip P is designed for aspirating 1 mL to 10 mL from beakers
  • Eppendorf Varitip S for 2.5 mL to 10 mL forms a system with the Varitip and is used for aspirating liquid from tall, narrow-necked vessels
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Valve
100 Valves
Product Title
Eppendorf™ Valve, For Varitip S > Valve, For Varitip S