Learn More
Eppendorf™ Valve, For Varitip S
Tailored to different vessels. Choose from a variety of Eppendorf Varitips to meet your needs.
Brand: Eppendorf™ 0030050541
890.00 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Eppendorf Varitip P is designed for aspirating 1 mL to 10 mL from beakers
- Eppendorf Varitip S for 2.5 mL to 10 mL forms a system with the Varitip and is used for aspirating liquid from tall, narrow-necked vessels
Specifications
Valve | |
100 Valves |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Eppendorf™ Valve, For Varitip S > Valve, For Varitip S
Spot an opportunity for improvement?Share a Content Correction