Learn More
Steris™ Vaprox Labware Detergent
Use the high-purity, specially formulated liquid sterilant Steris Corporation™ Vaprox Labware Detergent specially for healthcare environmental contamination control applications..
Brand: Steris™ PB006EUR
Additional Details : CAS Number : 7722-84-1 Weight : 6.00000kg Transport : UN number : 2014 Chem class : 5.1 Pack group : II
Descripción
- High purity
- Specially formulated liquid sterilant
- Developed for healthcare environmental contamination control applications.
Especificaciones
Disinfectant | |
US EPA (Registration No. 58779-4) | |
35% hydrogen peroxide |
Sterile | |
950 mL cartridge maximizes operator safety and designed so that lot number, expiration date and sterilant level are visible without removing the cartridge. | |
3 Cups/Case; 45 Cycles/Case |
Seguridad y manipulación
- Vaprox Hydrogen Peroxide Sterilant
Palabra de advertencia
- Danger
Clasificación
- Oxidising liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
Indicaciones de peligro
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
Consejos de prudencia
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to .