missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF Monoclonal Antibody (26503), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523719
This item is not returnable.
View return policy
Description
In ELISAs, this antibody shows approximately 10% cross-reactivity with recombinant mouse (rm) VEGF and rrVEGF and no cross-reactivity with rhVEGF-D. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
VEGF Monoclonal antibody specifically detects VEGF in Human, Non-human Primate samples. It is validated for ELISA, Neutralization, Western Blot.Specifications
VEGF | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P15692 | |
VEGFA | |
Sf 21-derived recombinant human VEGF165 Ala27-Arg191 | |
500 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG2b |
ELISA, Neutralization, Western Blot | |
26503 | |
Unconjugated | |
VEGFA | |
angiogenic factor; etID309976.16; eVEGF120; eVEGF164; human VEGF121 homolog; MGC70609; MVCD1; ovine vascular endothelial growth factor; RP1-261G23.1; unnamed protein product; VAS; vascular endothelial growth factor; vascular endothelial growth factor (VEGF) precursor; vascular endothelial growth factor 121 isoform; vascular endothelial growth factor 164; vascular endothelial growth factor 165b; vascular endothelial growth factor 182; vascular endothelial growth factor 182 isoform; vascular endothelial growth factor 188; vascular endothelial growth factor 188 isoform; vascular endothelial growth factor 189; vascular endothelial growth factor A; vascular endothelial growth factor A121; vascular endothelial growth factor A165; vascular endothelial growth factor Aa; Vascular endothelial growth factor A-A; vascular endothelial growth factor isoform 165; vascular permeability factor; Vasculotropin; VEGF; VEGF1; VEGF164; VEGF182; VEGF188; vegfa; VEGF-A; VEGFA165b; vegfaa; VEGF-A-A; VEGFMGC706 | |
Mouse | |
Protein A/G | |
RUO | |
7422 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |