Learn More
Thermo Scientific™ Versette™ Accessories
Use Thermo Scientific™ Versette™ Accessories with the Versette™ Automated Liquid Handling System.
Brand:  Thermo Scientific™ 650-08-9630SD
 View more versions of this product
Product Code. 15403780
Description
Description
Use Thermo Scientific™ Versette™ Accessories with the Versette™ Automated Liquid Handling System.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Serial Dilute Magazine
Serial dilute magazine 96/30μL (8/12) for use with short 30μL tips
96
30 μL