Learn More
Thermo Scientific™ Versette™ Accessories
Use Thermo Scientific™ Versette™ Accessories with the Versette™ Automated Liquid Handling System.
Brand:  Thermo Scientific™ 650-04-PUMP
 View more versions of this product
Product Code. 15493770
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Use Thermo Scientific™ Versette™ Accessories with the Versette™ Automated Liquid Handling System.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pump module; Optional accessory, used for tip washing/reagent replenishing; 12.7 kg (28 lbs.)
Pump Module