Learn More
Heidolph instruments Vial MonoBlock
Accommodate specifically sized reagent tubes at your StarFish magnetic stirrer workstation. Heidolph™ Vial MonoBlock, constructed of one piece, is ideal for experiments using the same vessel.
Brand: Heidolph instruments 505-80007-00
This item is not returnable.
View return policy
Description
- Accommodates one specific vial size.
- One-piece construction.
Specifications
MonoBlock | |
Starfish Workstation, MR-Hei Magnetic Stirrers | |
20 x 21 mm vials | |
115/240 V |