missing translation for 'onlineSavingsMsg'
Learn More

Vinblastine sulfate, 100.2%, MP Biomedicals™

Product Code. 11401821
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 143-67-9
Molecular Formula C46H60N4O13S
Molecular Weight (g/mol) 909.06
MDL Number MFCD08706468,MFCD00082457,MFCD08706468
InChI Key KDQAABAKXDWYSZ-JKDPCDLQSA-N
Synonym vinblastine sulfate
PubChem CID 131709225
IUPAC Name methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(13S,15S,17S)-17-ethyl-17-hydroxy-13-(methoxycarbonyl)-1,11-diazatetracyclo[13.3.1.0⁴,¹².0⁵,¹⁰]nonadeca-4(12),5,7,9-tetraen-13-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate; sulfuric acid
SMILES OS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(C1)CCC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC

Specifications

Color White to Yellow
Quantity 1 mg
Formula Weight 909.1
Physical Form Powder
Chemical Name or Material Vinblastine sulfate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'