missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinblastine sulfate, 100.2%, MP Biomedicals™
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Specifications
Specifications
Color | White to Yellow |
Quantity | 25 mg |
Formula Weight | 909.1 |
Physical Form | Powder |
Chemical Name or Material | Vinblastine sulfate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'