missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Vinyl Pre-Printed Chemical Labels
Brady™ Vinyl GHS/CLP Chemical Labels for BBP3x printers are self-adhesive and preprinted with red diamonds in a variety of layouts.
Brand: Brady™ 121645
This item is not returnable.
View return policy
Description
- BBP™31 and BBP™33 compatible labels
- Blank labels with pre-printed red diamonds allow printer to produce two-color GHS/CLP labels in one print pass
- Available in three sizes in the full product portfolio, each with six diamond layouts that match templates in MarkWare™Software
- Service temperature -20°C to 75°C; minimum application temperature 10°C
- Average outdoor durability 3-5 years
Specifications
BS5609:1986 Section 2 and section 3 | |
Acrylic, Permanent | |
Gloss | |
Pre-printed Chemical Label | |
1 | |
200/Pk. | |
-20°C to +75°C | |
155 x 105 mm | |
155.00 mm | |
0.1 kg |
1 Diamond | |
White, Red | |
Vinyl | |
Facility and Safety identification, Hazardous Substance identification | |
Up to 3 Years Outdoor Use | |
MarkWare | |
0.026 mm | |
105 mm | |
155 mm |