missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab Violet Red Bile Agar (VRB Agar)
Scharlab Violet Red Bile Agar is for the detection and enumeration of coliforms in milk and other dairy products.
Brand: Scharlab 01-164-500
This item is not returnable.
View return policy
Description
- APHA/COMPF/IDF-FIL/ISO
- Medium for the detection and enumeration of coliforms in milk and other dairy products
- According to APHA and ICMSF, FIL-IDF 73B and ISO 4832 and 5541-1 standards.
Specifications
ISO 4832, 5541-1 | |
Solid | |
Microbiology Media | |
Yeast Extract 3.000 g/L, Peptone 7.000 g/L, Bile Salts No. 3 1.500 g/L, Lactose 10.000 g/L, Sodium Chloride 5.000 g/L, Neutral Red 0.030 g/L, Crystal Violet 0.002 g/L, Agar 13.000 g/L | |
30°C ± 1.0°C | |
7.4 ±0.2 |
Red | |
Violet Red Bile Agar | |
Coliforms | |
Detection and Enumeration of Coliforms in Milk and Other Dairy Products | |
24 ±2 hr. | |
500 g |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Scharlab Violet Red Bile Agar (VRB Agar) > Violet Red Bile Agar; Form: Solid
Spot an opportunity for improvement?Share a Content Correction