Learn More
Viscosity Reference Standard Low Temperature Type N27B UKAS ISO 17025 / ISO 17034 Certified, 500 mL, Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified low temperature viscosity standards are for the calibration and verification of glass capillary viscometers and other viscosity measuring equipment where operating temperature is controlled precisely.
Brand:  Paragon Scientific N27B
Additional Details : Weight : 0.50000kg
Product Code. 16080270
  • 1080.00 SEK / 500mL
Estimated Shipment: 20-09-2024
to see stock.
Description
Description
  • Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Standard Pack Size 500mL, available also in 5L. Please contact sales@paragon-sci.com for other volumes
  • Fully traceable to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
  • Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    Special Offers
    Special Offers
    Nominal values quoted. Certified Values will vary from batch to batch.