missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Visocolor™ HE Colorimetric Test Kit
Provides the highest sensitivity and accuracy in visual analytics. Macherey-Nagel™ Visocolor™ HE Colorimetric Test Kits determine even the lowest limiting values using longer measuring tubes and larger sample volumes. Includes all reagents and testing equipment/supplies.
1315.00 SEK - 1340.00 SEK
Specifications
Description | Highly sensitive |
---|---|
Format | HE Colorimetric Test Kit |
Includes | Reagents and test equipment (e.g. glass tubes with screw caps, comparator block, color comparison disk) |
Quantity | 1 Each |
Type | Water analysis test kit |
Product Code | Brand | For Use With (Application) | Range | No. of Tests | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Range | No. of Tests | Price | Quantity & Availability | |||
10628061
|
Macherey-Nagel™
920055 |
Manganese determination | 0 to 0.50mg/L | 100 |
1315.00 SEK
100 tests |
Estimated Shipment: 18-09-2024
Log in to see stock. |
|||
10024260
|
Macherey-Nagel™
920040 |
Iron determination | 0 to 0.20mg/L | 300 |
1340.00 SEK
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
|||
Description
Visocolor™ HE test kits provide highly precise analysis via exact dosing of the single reagents as well as compensating for turbidity and color. The sensitivity of Visocolor™ HE is 10 to 100 times higher compared to other Visocolor tests.
- Visual evaluation
- Environment-friendly, without toxic reagents
- Economically priced
- Convenient handling
- Highest accuracy due to extremely narrow gradation
- Highest sensitivity down to 0.002mg/L due to longer measuring tubes
- Reagent bottles with clear dosing instructions
- Compensation of turbidities and colors
- Refill packs available
Specifications
Highly sensitive | |
Reagents and test equipment (e.g. glass tubes with screw caps, comparator block, color comparison disk) | |
Water analysis test kit |
HE Colorimetric Test Kit | |
1 Each |