Learn More
WTW™ VisoTurb™ 900-P IDS IR Turbidity Sensor
The WTW™ VisoTurb™ 900-P IDS IR Turbidity Sensor is designed for digital IDS turbidity measurement for field applications in surface waters and process samples.
Brand: WTW™ 600700
19621.64 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Digital IDS turbidity measurement with for mobile on-site applications in surface waters, pump tests and process samples
- For use with all MultiLine™ IDS portable meters and in the lab with the inoLab™ Multi 9310 IDS, 9620 IDS and 9630 IDS models
- Easy two or three-point calibration
- Suitable for multi-parameter measurements
- Robust and reliable
- Corrosion-resistant titanium housing, waterproof up to 10 bar
- Use with IDS wireless modules is recommended
Specifications
Digital Turbidity Sensor, Wireless Ready | |
MultiLine IDS Portable Meters, InoLab Multi 9310 IDS, 9620 IDS and 9630 IDS | |
Universal plug head | |
IDS wireless modules |
Product Content Correction
Product Title
WTW™ VisoTurb™ 900-P IDS IR Turbidity Sensor > Includes universal plug head
Spot an opportunity for improvement?Share a Content Correction