Learn More
Vistusertib, MedChemExpress
MedChemExpress Vistusertib (AZD2014) is an ATP competitive mTOR inhibitor with an IC50 of 2.81 nM. AZD2014 inhibits both mTORC1 and mTORC2 complexes.
Brand: Medchem Express HY-15247
Description
Vistusertib (AZD2014) is an ATP competitive mTOR inhibitor with an IC50 of 2.81 nM. AZD2014 inhibits both mTORC1 and mTORC2 complexes.Specifications
Vistusertib | |
Yellow | |
C25H30N6O3 | |
AZD2014 | |
O=C(NC)C1=CC=CC(C2=CC=C3C(N=C(N4[C@@H](C)COCC4)N=C3N5[C@@H](C)COCC5)=N2)=C1 | |
462.54 | |
Research | |
Cancer-Kinase/protease |
1009298-59-2 | |
Research | |
5 mg | |
DMSO : ≥ 50 mg/mL (108.10 mM) | |
462.54 | |
95.66% | |
Solid |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only