Learn More
Vitronectin Antibody (A18), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-23571PCP
Product Code. 18444652
  • 10995.00 SEK / 0.25mL
Explore more special offers
Description
Description
Vitronectin Monoclonal antibody specifically detects Vitronectin in Bovine, Equine, Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Western Blot (Negative)
Specifications
Specifications
Vitronectin
Monoclonal
PerCP
PBS
Mouse
Protein A or G purified
RUO
Primary
Bovine, Equine, Rabbit
Purified
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
A18
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Western Blot (Negative)
complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT
Lysed bovine corneal endothelial cells and extracellular matrix
0.25 mL
Immunology, Innate Immunity
7448
Store at 4C in the dark.
IgG1 κ