Learn More
Vitlab™ VITsafe™ Narrow-Mouth Wash Bottles with Imprint
Identify media quickly and easily with these color-coded wash bottles. VITsafe™ Narrow-Mouth Wash Bottles with Imprint feature a precise spray jet and optimized medium backflow through the continuous and finely drawn tip of the spray tube.
Brand: Vitlab™ 1433829
1439.25 SEK valid until 2024-12-31
Use promo code "23277" to get your promotional price.
Additional Details : Weight : 1.33200kg
Description
- Bottles made from LDPE (or PP for Acetone and MEK), with polypropylene spray tube
- VENT-CAP practically eliminates leakage and prevents almost all static overpressure
- Color-coded for easy visual identification of the media
- Material name in German, English, French and Spanish
- Chemical formula, CAS No., hazard pictogram, signal word
- Risk phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
Specifications
1000 mL | |
Screw Cap, Spray Tube | |
12 Pack | |
Directive (EC) No. 1272/2008 (GHS) | |
Narrow | |
Wash Bottle | |
92 mm |
Imprint: Acetone | |
LDPE | |
Red | |
Yes | |
215 mm | |
Polypropylene | |
Vented |