missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivacon™ 500, Sample Pack L (Low MWCO)

Vivacon™ 500 are centrifugal units with Hydrosart™ membranes for fast DNA concentration and buffer exchange of samples of 100–500μL. Sample Pack L is intended for use with samples with a low molecular weight cut off, and include 4 units each of the 2, 10 and 30 kDa sets.

Brand:  Sartorius VN01HL12

Product Code. 11750896

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Vivacon™ 500 is optimized for the concentration / recovery and buffer exchange applications of genomic DNA and peptides for samples up to 500μl initial volume. They are also the product of choice for peptide fractionation applications in this volume range. For optimal performance with highly diluted samples, e.g. forensic specimens, Vivacon™ 500 is equipped with patented Hydrosart™ regenerated cellulose membranes with extremely low unspecific binding properties and high flux.

High recoveries and excellent reproducibility are paired with convenience offered by volume graduation and molecular weight cut-offs printed on individual devices. Vivacon™ 500 features a horizontal membrane design optimal for DNA fractionation (primer removal) as well as peptide fractionation (Filter Aides Sample Preparation, FASP1). The possibility of a reverse spin after sample processing assures complete concentrate recovery which is especially important when working with low sample concentrations.

  • Concentration of up to 0.5 ml genomic DNA or peptide samples
  • Low binding material
  • Reverse spin possibility
  • MWCO printed on device
  • Complete and highly reproducible concentrate recovery
TRUSTED_SUSTAINABILITY
Specifications

Specifications

None
5 μL
Hydrosart
0.1 to 0.5 mL
DNA, Peptide
0.1 to 0.5 mL
4 each of 2, 10 and 30 kDa units
Polycarbonate Concentrator Body/Sleeve, Polypropylene Filtrate Vessel
30 to 100 kDa
Centrifugal UltraFiltration
Ultrafiltration Devices
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers