missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS41 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13869-1-AP-150UL
This item is not returnable.
View return policy
Description
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human ortholog of yeast Vps41 protein which is also conserved in Drosophila, tomato, and Arabidopsis. Expression studies in yeast and human indicate that this protein may be involved in the formation and fusion of transport vesicles from the Golgi. Several transcript variants encoding different isoforms have been described for this gene, however, the full-length nature of not all is known.Specifications
VPS41 | |
Polyclonal | |
Unconjugated | |
VPS41 | |
HVPS41, hVps41p, HVSP41, S53, VPS41 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27072 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P49754 | |
VPS41 | |
VPS41 Fusion Protein Ag4760 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |