Learn More
vwf Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27186-1-AP-150UL
 View more versions of this product
Product Code. 16895455
  • 4721.67 SEK / 150µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
Von Willebrand factor (vWf) is a series of self-aggregated structures, all derived from common glycoprotein subunits, synthesized in endothelial cells and megakaryocytes. The molecular mass of the monomeric vWf unit is 225 kDa. vWf binds to circulating factor VIIIc (antihemophilic factor), hereby stabilizing the latter thus being an important factor in the normal primary haemostasis.
Specifications
Specifications
vwf
Polyclonal
Unconjugated
VWF
F8VWF, von Willebrand antigen II, von Willebrand factor, VWD, vwf
Rabbit
Antigen Affinity Chromatography
RUO
7450
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P04275
VWF
vwf Fusion Protein Ag25578
150 μL