Learn More
Conair™ Waring™ Mini-sample Containers for Blender
Stainless steel containers
6030.00 SEK
Specifications
Product Type Mini Sample Container
For Use With (Equipment) Waring™ 1 L blender
View More Specs
Products 1
Product Code Brand Volume Range (Metric) Price Quantity & Availability  
Product Code Brand Volume Range (Metric) Price Quantity & Availability  
11964938
View Documents
Conair™
MC3
50 to 250 mL
6030.00 SEK
Each
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
Description
Description
Stainless-steel blending assemblies are for use with 1L (34 oz.) capacity blenders (nominal capacity; actual capacity depends on container used).
  • Snap-fit cover
  • Cloverleaf design
Specifications
Specifications
Mini Sample Container
Waring™ 1 L blender