Learn More
Waring™ Timer Blender
Brand:  Waring™ 21/8010EG
Product Code. 12879385
  • 15935.00 SEK / Each
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Description
Multi-speed blenders, with a choice of either glass or stainless steel containers, and the unique clover shape for perfect blending action. Waring blenders are designed for frequent use.
Specifications
Specifications
Timer Blender
1 L
Timer
Certificates
View accessory products
Product Title
Waring™ Timer Blender > Capacity: 1L