Learn More
Lenz Laborglasinstrumente™ Wash Bottles
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Description
- Wash bottles
- Upright piston according to DIN 12348
- Made of DURAN™
Specifications
Capacity (Metric) | 500 mL |
Material | Borosilicate Glass |
Certifications/Compliance | DIN 12348 |