Learn More
BRAND™ Watch Glass
Watch Glass constructed of soda-lime glass with ground edges.
Brand: BRAND™ 150060
636.56 SEK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
- Soda-lime glass
- Ground edges
- Stress-reduced to minimize risk of shattering
Specifications
Stress reduced edges | |
Soda Lime Glass | |
Ground | |
120 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Product Content Correction
Product Title
BRAND™ Watch Glass > Diameter: 120 mm
Spot an opportunity for improvement?Share a Content Correction